Assay Details
Target Gene Details
Entrez Gene ID: | 107035 |
Gene Name: | F-box protein 38 |
Gene Aliases: |
6030410I24Rik, AU044865, AW214031, Moka, SP329 |
Location: |
Chr.18:62504059-62548858 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fbxo38 | NM_134136.3 | 2 | 218 | NP_598897.2 |
AK031096.1 | 2 | 168 | BAC27250.1 | |
AK031347.1 | 2 | 149 | BAC27358.1 | |
AK089046.1 | 2 | 190 | BAC40718.1 | |
AY267463.1 | 2 | 151 | AAP31028.1 | |
BC056348.1 | 2 | 170 | AAH56348.1 | |
BY131284.1 | 2 | 218 |