Assay Details
Target Gene Details
Entrez Gene ID: | 24010 |
Gene Name: | IK cytokine |
Gene Aliases: |
MuRED |
Location: |
Chr.18:36744656-36757639 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ik | NM_011879.2 | NP_036009.1 | ||
AJ006130.1 | CAA06880.1 | |||
AK029644.1 | BAC26544.1 | |||
AK050625.1 | BAC34349.1 | |||
AK082356.1 | ||||
AK087976.1 | BAC40070.1 | |||
AK088908.1 | ||||
AK163766.1 | BAE37486.1 | |||
AK167238.1 | BAE39360.1 | |||
AK203563.1 | ||||
BC014739.1 | AAH14739.1 | |||
BC026569.1 | AAH26569.1 | |||
BC029671.1 | AAH29671.1 | |||
BC029694.1 | AAH29694.1 | |||
BC033356.1 | AAH33356.1 | |||
BG084120.2 |
More Information
Set Membership: |
Intragenic Non-exonic |