Assay Details
Target Gene Details
Entrez Gene ID: | 24010 |
Gene Name: | IK cytokine |
Gene Aliases: |
MuRED |
Location: |
Chr.18:36744656-36757639 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ik | NM_011879.2 | 20 | 1917 | NP_036009.1 |
AK082356.1 | 20 | 1850 | ||
AK087976.1 | 20 | 1863 | BAC40070.1 | |
BC014739.1 | 20 | 1848 | AAH14739.1 | |
BC026569.1 | 20 | 1848 | AAH26569.1 | |
BC029671.1 | 20 | 1860 | AAH29671.1 | |
BC029694.1 | 20 | 1857 | AAH29694.1 | |
BC033356.1 | 20 | 1844 | AAH33356.1 | |
BM211698.2 | 1 | 80 |
More Information
Set Membership: |
Intragenic Exonic |