Assay Details
Target Gene Details
Entrez Gene ID: | 58523 |
Gene Name: | elongator acetyltransferase complex subunit 2 |
Gene Aliases: |
AU023723, Epl2, StIP1, Statip1 |
Location: |
Chr.18:24603923-24638830 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Elp2 | NM_021448.2 | NP_067423.2 | ||
AF291064.1 | AAG01032.1 | |||
AK033246.1 | BAC28210.1 | |||
AK034457.1 | BAC28716.1 | |||
AK044699.1 | BAC32039.1 | |||
AK050960.1 | BAC34477.1 | |||
AK167725.1 | BAE39766.1 | |||
AK167853.1 | BAE39871.1 | |||
AK168370.1 | BAE40302.1 | |||
BC015284.1 | AAH15284.1 | |||
BC051943.1 | AAH51943.1 |