Assay Details
Target Gene Details
Entrez Gene ID: | 20730 |
Gene Name: | serine peptidase inhibitor, Kazal type 1 |
Gene Aliases: |
Spink3, p12 |
Location: |
Chr.18:43728069-43737237 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spink1 | NM_009258.5 | 1 | 74 | NP_033284.1 |
AK007841.1 | BAB25298.1 | |||
AK007985.1 | BAB25389.1 | |||
AV048854.1 | ||||
BC086887.1 | AAH86887.1 | |||
CA481615.1 | 1 | 111 | ||
X06342.1 | CAA29648.1 |
More Information
Set Membership: |
Intragenic Exonic |