Assay Details
Target Gene Details
Entrez Gene ID: | 15562 |
Gene Name: | 5 hydroxytryptamine (serotonin) receptor 4 |
Gene Aliases: |
5-HT-4, 5-HT4, 5-HT<4L>, 5HTR4 |
Location: |
Chr.18:62322160-62498026 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Htr4 | NM_008313.4 | 4 | 421 | NP_032339.2 |
AJ011369.1 | 3 | 211 | CAA09598.1 | |
AK141587.1 | 4 | 365 | BAE24750.1 | |
AK141625.1 | 4 | 422 | BAE24773.1 | |
JN418927.1 | 4 | 352 | ||
JN418928.1 | 4 | 338 | ||
JN418929.1 | 3 | 211 | ||
JN418930.1 | 3 | 227 | ||
Y09585.1 | 3 | 211 | CAA70773.1 | |
Y09587.1 | 3 | 211 | CAA70775.1 | |
Y09588.1 | 3 | 211 | CAA70776.1 |
More Information
Set Membership: |
Intragenic Exonic |