Assay Details
Target Gene Details
Entrez Gene ID: | 81500 |
Gene Name: | endoplasmic reticulum chaperone SIL1 homolog (S. cerevisiae) |
Gene Aliases: |
1810057E01Rik, AI042831, wz |
Location: |
Chr.18:35266396-35499052 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sil1 | NM_030749.2 | NP_109674.2 | ||
AJ297884.1 | CAC17789.1 | |||
AK170415.1 | BAE41781.1 | |||
AK206841.1 | ||||
AK208966.1 | ||||
AK217356.1 | ||||
BC016119.1 | AAH16119.1 | |||
BC016466.1 | AAH16466.1 | |||
BX512650.1 | ||||
CO428760.1 |