Assay Details
Target Gene Details
Entrez Gene ID: | 81500 |
Gene Name: | endoplasmic reticulum chaperone SIL1 homolog (S. cerevisiae) |
Gene Aliases: |
1810057E01Rik, AI042831, wz |
Location: |
Chr.18:35266396-35499052 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sil1 | NM_030749.2 | 6 | 744 | NP_109674.2 |
AJ297884.1 | 6 | 685 | CAC17789.1 | |
AK170415.1 | 6 | 697 | BAE41781.1 | |
AK192274.1 | 1 | 64 | ||
AK219586.1 | 1 | 64 | ||
BC016119.1 | 6 | 721 | AAH16119.1 | |
BC016466.1 | 6 | 707 | AAH16466.1 | |
CO428760.1 | 3 | 287 | ||
CX219879.1 | 6 | 744 |