Assay Details
Target Gene Details
Entrez Gene ID: | 15562 |
Gene Name: | 5 hydroxytryptamine (serotonin) receptor 4 |
Gene Aliases: |
5-HT-4, 5-HT4, 5-HT<4L>, 5HTR4 |
Location: |
Chr.18:62322160-62498026 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Htr4 | NM_008313.4 | 1 | 78 | NP_032339.2 |
AK141587.1 | BAE24750.1 | |||
AK141625.1 | 1 | 79 | BAE24773.1 | |
JN418927.1 |
More Information
Set Membership: |
Intragenic Exonic |