Assay Details
Target Gene Details
Entrez Gene ID: | 108112 |
Gene Name: | eukaryotic translation initiation factor 4E binding protein 3 |
Gene Aliases: |
4e-bp3 |
Location: |
Chr.18:36664060-36666324 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif4ebp3 | NM_201256.4 | 2 | 291 | NP_957708.1 |
AY226182.1 | 2 | 169 | AAO73448.1 | |
BC061242.1 | 2 | 291 | AAH61242.1 | |
BC111107.1 | 2 | 231 | AAI11108.1 |
More Information
Set Membership: |
Intragenic Exonic |