Assay Details
Target Gene Details
Entrez Gene ID: | 52563 |
Gene Name: | CDC23 cell division cycle 23 |
Gene Aliases: |
6030435O18, D18Ertd243e |
Location: |
Chr.18:34630946-34651745 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdc23 | NM_178347.4 | NP_848124.1 | ||
AK031459.1 | BAC27415.1 | |||
AK038523.1 | BAC30026.1 | |||
AK047789.1 | ||||
AK048246.1 | BAC33283.1 | |||
AK052567.1 | ||||
AK075948.1 | ||||
AK079681.1 | ||||
AK081461.1 | BAC38224.1 | |||
AK169677.1 | BAE41298.1 | |||
AK201802.1 | ||||
AK217936.1 | ||||
AK219876.1 | ||||
BC059013.1 | AAH59013.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |