Assay Details
Target Gene Details
Entrez Gene ID: | 319888 |
Gene Name: | O-acyltransferase like |
Gene Aliases: |
5330437I02Rik, Acyl3 |
Location: |
Chr.18:65697370-65751624 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oacyl | NM_177028.3 | 5 | 677 | NP_796002.1 |
AK030604.1 | 5 | 677 | BAC27041.1 | |
AK030615.1 | 3 | 457 | BAC27048.1 | |
BC141265.1 | 5 | 674 |
More Information
Set Membership: |
Intragenic Exonic |