Assay Details
Target Gene Details
Entrez Gene ID: | 224661 |
Gene Name: | solute carrier family 26, member 8 |
Gene Aliases: |
- |
Location: |
Chr.17:28637779-28691237 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc26a8 | NM_001290320.1 | 6 | 958 | NP_001277249.1 |
BC027076.1 | 6 | 958 | AAH27076.1 |
More Information
Set Membership: |
Intragenic Exonic |