Assay Details
Target Gene Details
Entrez Gene ID: | 66972 |
Gene Name: | solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23 |
Gene Aliases: |
2310067G05Rik, SCaMC-3 |
Location: |
Chr.17:57043711-57059880 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a23 | NM_025877.4 | 5 | 708 | NP_080153.2 |
AK191165.1 | 2 | 75 | ||
AK199713.1 | 2 | 75 | ||
AK207800.1 | 2 | 85 | ||
AK210726.1 | 2 | 80 | ||
AK210735.1 | 2 | 77 | ||
BC072660.1 | 5 | 704 | AAH72660.1 |
More Information
Set Membership: |
Intragenic Exonic |