Assay Details
Target Gene Details
Entrez Gene ID: | 60532 |
Gene Name: | Wilms tumour 1-associating protein |
Gene Aliases: |
2810408K05Rik, 9430038B09Rik |
Location: |
Chr.17:12966798-12992808 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wtap | NM_001113532.1 | NP_001107004.1 | ||
NM_001113533.1 | NP_001107005.1 | |||
NM_175394.2 | NP_780603.1 | |||
AK031965.1 | ||||
AK076111.1 | BAC36191.1 | |||
AK156334.1 | BAE33680.1 | |||
BC046416.1 | AAH46416.1 | |||
BC093504.1 | AAH93504.1 | |||
BY016042.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |