Assay Details
Target Gene Details
Entrez Gene ID: | 60532 |
Gene Name: | Wilms tumour 1-associating protein |
Gene Aliases: |
2810408K05Rik, 9430038B09Rik |
Location: |
Chr.17:12966798-12992808 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wtap | NM_175394.2 | 1 | 39 | NP_780603.1 |
AK031965.1 | 1 | 47 | ||
AK076111.1 | BAC36191.1 |