Assay Details
Target Gene Details
Entrez Gene ID: | 20519 |
Gene Name: | solute carrier family 22 (organic cation transporter), member 3 |
Gene Aliases: |
EMT, Oct3, Orct3, Slca22a3 |
Location: |
Chr.17:12419972-12507704 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc22a3 | NM_011395.2 | NP_035525.1 | ||
AF078750.1 | AAD20978.1 | |||
AF082566.1 | AAD53007.1 | |||
AK143988.1 | BAE25648.1 | |||
BC137661.1 | ||||
BC137662.1 |
More Information
Set Membership: |
Intragenic Non-exonic |