Assay Details
Target Gene Details
Entrez Gene ID: | 67681 |
Gene Name: | mitochondrial ribosomal protein L18 |
Gene Aliases: |
1010001C05Rik, C79879, HSPC071, MRP-L18 |
Location: |
Chr.17:12911345-12916478 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrpl18 |
Target Gene Details
Entrez Gene ID: | 21454 |
Gene Name: | t-complex protein 1 |
Gene Aliases: |
AI528772, CCT, Cct1, Ccta, TRic, Tcp-1, Tp63, c-cpn, p63 |
Location: |
Chr.17:12916329-12925067 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tcp1 | NM_001290712.1 | 1 | 182 | NP_001277641.1 |
NM_013686.4 | 1 | 182 | NP_038714.2 | |
AK149611.1 | BAE28989.1 | |||
AK149755.1 | BAE29063.1 | |||
AK150666.1 | 1 | 62 | ||
AK150744.1 | 1 | 62 | ||
AK151068.1 | BAE30084.1 | |||
AK151445.1 | 1 | 72 | BAE30407.1 | |
AK151903.1 | 1 | 62 | ||
AK152633.1 | 1 | 62 | ||
AK152641.1 | BAE31381.1 | |||
AK152759.1 | ||||
AK153037.1 | ||||
AK153430.1 | 1 | 72 | BAE31988.1 | |
AK165665.1 | BAE38327.1 | |||
AK166828.1 | BAE39052.1 | |||
AK166966.1 | 1 | 70 | BAE39149.1 | |
AK167529.1 | BAE39599.1 | |||
BC003809.1 | AAH03809.1 | |||
CN725473.1 | 1 | 182 | ||
D90344.1 | BAA14356.1 | |||
M12899.1 | AAA40338.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |