Assay Details
Target Gene Details
Entrez Gene ID: | 94224 |
Gene Name: | steroid 5 alpha-reductase 2 |
Gene Aliases: |
5ART2, S5AR 2 |
Location: |
Chr.17:74017706-74047916 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srd5a2 | NM_053188.2 | NP_444418.1 | ||
AB049456.1 | BAB40179.1 | |||
AK138946.1 | BAE23830.1 | |||
BC125510.1 |
More Information
Set Membership: |
Intragenic Non-exonic |