Assay Details
Target Gene Details
Entrez Gene ID: | 12558 |
Gene Name: | cadherin 2 |
Gene Aliases: |
CDHN, N-cadherin, Ncad |
Location: |
Chr.18:16588877-16809049 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh2 | NM_007664.4 | 13 | 2744 | NP_031690.3 |
AB008811.1 | 13 | 2421 | BAA23549.1 | |
AK032826.1 | 13 | 2184 | BAC28044.1 | |
AK146981.1 | 13 | 2941 | BAE27584.1 | |
BC022107.1 | 13 | 2175 | AAH22107.1 | |
M31131.1 | 13 | 2421 | AAA37353.1 |
More Information
Set Membership: |
Intragenic Exonic |