Assay Details
Target Gene Details
Entrez Gene ID: | 12385 |
Gene Name: | catenin (cadherin associated protein), alpha 1 |
Gene Aliases: |
2010010M04Rik, AA517462, AI988031, Catna1 |
Location: |
Chr.18:35118912-35254775 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctnna1 | NM_009818.1 | 14 | 2024 | NP_033948.1 |
AK004623.1 | 14 | 1982 | BAB23418.1 | |
AK036752.1 | 14 | 2049 | ||
AK046513.1 | 8 | 1383 | ||
AK148496.1 | 14 | 2021 | BAE28588.1 | |
AK149721.1 | 14 | 2039 | ||
AK149738.1 | 14 | 2039 | ||
AK159339.1 | 14 | 2039 | BAE35001.1 | |
AK168635.1 | 14 | 2049 | BAE40495.1 | |
BC048163.1 | 14 | 2012 | AAH48163.1 | |
BC068316.1 | 14 | 2016 | AAH68316.1 | |
BC105576.1 | 14 | 2004 | ||
D90362.1 | 14 | 2015 | BAA14376.1 | |
X59990.1 | 14 | 2024 | CAA42607.1 |
More Information
Set Membership: |
Intragenic Exonic |