Assay Details
Target Gene Details
Entrez Gene ID: | 72238 |
Gene Name: | TBC1 domain family, member 5 |
Gene Aliases: |
1600014N05Rik |
Location: |
Chr.17:50733124-51180198 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tbc1d5 | NM_001285991.1 | 19 | 2209 | NP_001272920.1 |
NM_001285993.1 | 18 | 2110 | NP_001272922.1 | |
NM_028162.4 | 18 | 2143 | NP_082438.3 | |
AK154975.1 | 19 | 2169 | BAE32965.1 | |
AK163366.1 | 18 | 2173 | ||
AK220251.1 | 18 | 2110 | BAD90176.1 | |
BC043113.1 | 18 | 1959 | AAH43113.1 | |
BC098328.1 | 13 | 1489 | AAH98328.1 |
More Information
Set Membership: |
Intragenic Exonic |