Assay Details
Target Gene Details
Entrez Gene ID: | 224674 |
Gene Name: | solute carrier family 37 (glycerol-3-phosphate transporter), member 1 |
Gene Aliases: |
G3PP |
Location: |
Chr.17:31294798-31350709 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc37a1 | NM_001242427.1 | NP_001229356.1 | ||
NM_153062.2 | NP_694702.1 | |||
AK033656.1 | BAC28411.1 | |||
AK143636.1 | BAE25474.1 | |||
AK153706.1 | BAE32150.1 | |||
AK163990.1 | BAE37574.1 | |||
BC027294.1 | AAH27294.1 |
More Information
Set Membership: |
Intragenic Non-exonic |