Assay Details
Target Gene Details
Entrez Gene ID: | 19274 |
Gene Name: | protein tyrosine phosphatase, receptor type, M |
Gene Aliases: |
RPTPmu, mKIAA4044 |
Location: |
Chr.17:66666848-67354541 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprm | NM_008984.2 | NP_033010.2 | ||
AK034753.1 | BAE43289.1 | |||
AK129005.1 | ||||
AK164246.1 | BAE37702.1 | |||
AK220171.1 | BAD90356.1 | |||
BC079621.1 | AAH79621.1 | |||
X58287.1 | CAA41225.1 |
More Information
Set Membership: |
Intragenic Non-exonic |