Assay Details
Target Gene Details
Entrez Gene ID: | 19274 |
Gene Name: | protein tyrosine phosphatase, receptor type, M |
Gene Aliases: |
RPTPmu, mKIAA4044 |
Location: |
Chr.17:66666848-67354541 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprm | NM_008984.2 | 26 | 4052 | NP_033010.2 |
AK034753.1 | 4 | 598 | BAE43289.1 | |
AK129005.1 | 24 | 3419 | ||
AK164246.1 | 4 | 464 | BAE37702.1 | |
AK220171.1 | 24 | 3323 | BAD90356.1 | |
BC079621.1 | 28 | 3768 | AAH79621.1 | |
X58287.1 | 26 | 3977 | CAA41225.1 |
More Information
Set Membership: |
Intragenic Exonic |