Assay Details
Target Gene Details
Entrez Gene ID: | 68816 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 1 |
Gene Aliases: |
1110060O10Rik, AI327391, Cypl1 |
Location: |
Chr.17:29250835-29263971 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppil1 | NM_026845.4 | 2 | 184 | NP_081121.1 |
AK004331.1 | 2 | 185 | BAB23265.1 | |
AK150643.1 | 2 | 161 | BAE29731.1 | |
AK151572.1 | 2 | 161 | BAE30513.1 | |
AK156809.1 | 2 | 185 | BAE33862.1 | |
AK217140.1 | ||||
BC058369.1 | 2 | 143 | AAH58369.1 | |
BC147647.1 | 2 | 269 | ||
BC147649.1 | 2 | 269 |
More Information
Set Membership: |
Intragenic Exonic |