Assay Details
Target Gene Details
Entrez Gene ID: | 74116 |
Gene Name: | peptidase inhibitor 16 |
Gene Aliases: |
1200009H11Rik, Cripi, PI-16 |
Location: |
Chr.17:29318796-29328903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pi16 | NM_023734.3 | NP_076223.3 | ||
AI587910.1 | ||||
AK004672.1 | ||||
AK157854.1 | ||||
AK158339.1 | ||||
BC039124.1 | AAH39124.1 | |||
BC118026.1 |
More Information
Set Membership: |
Intragenic Non-exonic |