Assay Details
Target Gene Details
Entrez Gene ID: | 56716 |
Gene Name: | MTOR associated protein, LST8 homolog (S. cerevisiae) |
Gene Aliases: |
0610033N12Rik, AA409454, AI505104, AI851821, Gbl |
Location: |
Chr.17:24473550-24479114 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mlst8 | NM_001252463.1 | 8 | 923 | NP_001239392.1 |
NM_001252464.1 | 8 | 898 | NP_001239393.1 | |
NM_001252465.1 | 6 | 707 | NP_001239394.1 | |
NM_019988.5 | 8 | 918 | NP_064372.2 | |
AF237676.1 | 8 | 849 | AAF37719.1 | |
AK002751.1 | 8 | 884 | BAB22328.1 | |
AK038515.1 | 7 | 1416 | BAC30024.1 | |
AK040100.1 | 8 | 919 | BAC30510.1 | |
AK048102.1 | 8 | 882 | BAC33243.1 | |
AK077680.1 | 8 | 889 | BAC36952.1 | |
AK083731.1 | 8 | 859 | BAC39006.1 | |
AK088951.1 | 8 | 892 | ||
BC015279.1 | 8 | 877 | AAH15279.1 | |
BM947104.1 | 6 | 689 |