Assay Details
Target Gene Details
Entrez Gene ID: | 19274 |
Gene Name: | protein tyrosine phosphatase, receptor type, M |
Gene Aliases: |
RPTPmu, mKIAA4044 |
Location: |
Chr.17:66666848-67354541 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprm | NM_008984.2 | 10 | 2040 | NP_033010.2 |
AK039261.1 | 1 | 173 | ||
AK129005.1 | 8 | 1407 | ||
AK220171.1 | 8 | 1311 | BAD90356.1 | |
BC058614.1 | 10 | 2050 | AAH58614.1 | |
BC079621.1 | 10 | 1654 | AAH79621.1 | |
X58287.1 | 10 | 1965 | CAA41225.1 |
More Information
Set Membership: |
Intragenic Exonic |