Assay Details
Target Gene Details
Entrez Gene ID: | 76498 |
Gene Name: | progestin and adipoQ receptor family member IV |
Gene Aliases: |
1500004C10Rik |
Location: |
Chr.17:23736186-23740898 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Paqr4 | NM_023824.3 | 2 | 544 | NP_076313.1 |
AB041564.1 | 2 | 465 | BAA95048.1 | |
AK005142.1 | 2 | 567 | BAB23839.1 | |
AK033625.1 | 2 | 562 | BAC28398.1 | |
AK044092.1 | 2 | 364 | BAC31770.1 | |
AK044486.1 | 2 | 567 | BAC31947.1 | |
AK152676.1 | 2 | 543 | BAE31408.1 | |
AY424293.1 | 2 | 495 | AAR08381.1 | |
BC011185.1 | 2 | 544 | AAH11185.1 | |
BC026921.1 | 2 | 495 | AAH26921.1 |
More Information
Set Membership: |
Intragenic Exonic |