Assay Details
Target Gene Details
Entrez Gene ID: | 14651 |
Gene Name: | hydroxyacyl glutathione hydrolase |
Gene Aliases: |
BC019817, Glo-2, Glo2, Rsp29 |
Location: |
Chr.17:24840079-24864450 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hagh | NM_001159626.1 | NP_001153098.1 | ||
NM_024284.2 | NP_077246.2 | |||
AK147022.1 | BAE27615.1 | |||
AK150557.1 | BAE29657.1 | |||
AK151231.1 | BAE30223.1 | |||
AK152742.1 | BAE31462.1 | |||
AK165777.1 | BAE38377.1 | |||
BC004749.1 | AAH04749.1 | |||
BC019817.1 | AAH19817.1 |