Assay Details
Target Gene Details
Entrez Gene ID: | 16004 |
Gene Name: | insulin-like growth factor 2 receptor |
Gene Aliases: |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
Location: |
Chr.17:12682406-12769706 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igf2r | NM_010515.2 | 28 | 4108 | NP_034645.2 |
AK088699.1 | 3 | 334 | BAC40512.1 | |
BC150817.1 | 28 | 4019 | ||
BC171973.1 | 28 | 4019 | ||
CA324933.1 | ||||
CN461279.1 | 2 | 157 | ||
U04710.1 | 28 | 4064 | AAA19568.1 |
More Information
Set Membership: |
Intragenic Exonic |