Assay Details
Target Gene Details
Entrez Gene ID: | 67993 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 12 |
Gene Aliases: |
0610016O18Rik |
Location: |
Chr.17:59000107-59013367 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nudt12 | NM_026497.2 | 4 | 992 | NP_080773.1 |
AK002641.1 | 4 | 993 | BAB22253.1 | |
AK083846.1 | 4 | 950 | ||
AK147867.1 | 4 | 946 | BAE28190.1 | |
BC057657.1 | 4 | 946 | AAH57657.1 |
More Information
Set Membership: |
Intragenic Exonic |