Assay Details
Target Gene Details
Entrez Gene ID: | 74142 |
Gene Name: | lon peptidase 1, mitochondrial |
Gene Aliases: |
1200017E13Rik, LON, Prss15 |
Location: |
Chr.17:56614297-56626903 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lonp1 | NM_028782.2 | 3 | 571 | NP_083058.2 |
AK004820.1 | 3 | 555 | BAB23591.1 | |
AK131910.1 | 3 | 557 | BAE20868.1 | |
AK157474.1 | 3 | 550 | BAE34094.1 | |
AK159302.1 | 3 | 556 | BAE34972.1 | |
AK160071.1 | 3 | 556 | BAE35606.1 | |
AK160076.1 | 3 | 557 | ||
AK161983.1 | 3 | 556 | BAE36666.1 | |
AK177008.1 | ||||
AY190302.1 | 3 | 541 | AAN85210.1 |
More Information
Set Membership: |
Intragenic Exonic |