Assay Details
Target Gene Details
Entrez Gene ID: | 74142 |
Gene Name: | lon peptidase 1, mitochondrial |
Gene Aliases: |
1200017E13Rik, LON, Prss15 |
Location: |
Chr.17:56614297-56626903 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lonp1 | NM_028782.2 | 15 | 2241 | NP_083058.2 |
AK004820.1 | 15 | 2225 | BAB23591.1 | |
AK131910.1 | 15 | 2227 | BAE20868.1 | |
AK157474.1 | 15 | 2220 | BAE34094.1 | |
AK159302.1 | 15 | 2226 | BAE34972.1 | |
AK160071.1 | 15 | 2226 | BAE35606.1 | |
AK160076.1 | 15 | 2227 | ||
AK161983.1 | 15 | 2226 | BAE36666.1 | |
AY190302.1 | 15 | 2211 | AAN85210.1 | |
BC092212.1 | 6 | 608 | AAH92212.1 |
More Information
Set Membership: |
Intragenic Exonic |