Assay Details
Target Gene Details
Entrez Gene ID: | 106707 |
Gene Name: | RNA pseudouridylate synthase domain containing 1 |
Gene Aliases: |
2310051D06Rik, AU019540, Rlucl |
Location: |
Chr.17:25727703-25731464 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpusd1 | NM_028009.3 | NP_082285.1 | ||
AK009926.1 | ||||
AK049334.1 | BAC33690.1 | |||
BC018186.1 | AAH18186.1 |
More Information
Set Membership: |
Intragenic Non-exonic |