Assay Details
Target Gene Details
Entrez Gene ID: | 74142 |
Gene Name: | lon peptidase 1, mitochondrial |
Gene Aliases: |
1200017E13Rik, LON, Prss15 |
Location: |
Chr.17:56614297-56626903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lonp1 | NM_028782.2 | NP_083058.2 | ||
AK004820.1 | BAB23591.1 | |||
AK131910.1 | BAE20868.1 | |||
AK157474.1 | BAE34094.1 | |||
AK159302.1 | BAE34972.1 | |||
AK160071.1 | BAE35606.1 | |||
AK160076.1 | ||||
AK161983.1 | BAE36666.1 | |||
AK203371.1 | ||||
AY190302.1 | AAN85210.1 | |||
BC092212.1 | AAH92212.1 |
More Information
Set Membership: |
Intragenic Non-exonic |