Assay Details
Target Gene Details
Entrez Gene ID: | 12488 |
Gene Name: | CD2-associated protein |
Gene Aliases: |
AL024079, C78928, METS-1, Mets1 |
Location: |
Chr.17:42749098-42876712 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd2ap | NM_009847.3 | NP_033977.3 | ||
AF077003.1 | AAC36099.1 | |||
AF149092.1 | AAF73150.1 | |||
AJ459109.1 | CAD30510.1 | |||
AK045218.1 | ||||
AK046750.1 | ||||
BB643447.1 | ||||
BC138374.1 | ||||
BC157901.1 |