Assay Details
Target Gene Details
Entrez Gene ID: | 106565 |
Gene Name: | delta-like 2 homolog (Drosophila) |
Gene Aliases: |
AI413481, DLK-2, Egfl9 |
Location: |
Chr.17:46297248-46303271 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dlk2 | NM_001286028.1 | NP_001272957.1 | ||
BC118057.1 | ||||
BC122518.1 | ||||
BY271273.1 | 1 | 55 |