Assay Details
Target Gene Details
Entrez Gene ID: | 14229 |
Gene Name: | FK506 binding protein 5 |
Gene Aliases: |
D17Ertd592e, Dit1, FKBP-5, FKBP51 |
Location: |
Chr.17:28398753-28486149 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fkbp5 | NM_010220.4 | 9 | 1107 | NP_034350.1 |
AK169795.1 | 9 | 1048 | BAE41371.1 | |
BC015260.1 | 9 | 1046 | AAH15260.1 | |
BU613862.1 | ||||
CF949740.1 | 2 | 136 | ||
CT010372.1 | 8 | 888 | CAJ18579.1 | |
U16959.1 | 9 | 1107 | AAA86983.1 | |
U36220.1 | 9 | 1035 | AAA89162.1 |
More Information
Set Membership: |
Intragenic Exonic |