Assay Details
Target Gene Details
Entrez Gene ID: | 224794 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 4 |
Gene Aliases: |
4933413N07Rik, AA986363, AI195364, E-NPP 4, Gm90, mKIAA0879 |
Location: |
Chr.17:44096307-44105924 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Enpp4 | NM_199016.2 | NP_950181.2 | ||
AK090098.1 | BAC41090.1 | |||
AK157622.1 | BAE34135.1 | |||
AK173048.1 | BAD32326.1 | |||
BC027749.1 | ||||
CJ182752.1 |