Assay Details
Target Gene Details
Entrez Gene ID: | 12263 |
Gene Name: | complement component 2 (within H-2S) |
Gene Aliases: |
- |
Location: |
Chr.17:34862602-34882100 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C2 | NM_013484.2 | 16 | 2101 | NP_038512.2 |
AK146812.1 | 16 | 2102 | BAE27452.1 | |
AK190654.1 | ||||
AK194662.1 | ||||
AK203677.1 | ||||
AK203686.1 | ||||
AK206586.1 | ||||
AK207402.1 | ||||
AK213705.1 | ||||
BC011086.1 | 16 | 2064 | AAH11086.1 | |
M16271.1 | AAA39562.1 | |||
M57891.1 | 16 | 2048 | AAA63294.1 |