Assay Details
Target Gene Details
Entrez Gene ID: | 225020 |
Gene Name: | fasciculation and elongation protein zeta 2 (zygin II) |
Gene Aliases: |
9030616F16, D17Ertd315e |
Location: |
Chr.17:78369212-78418152 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fez2 | NM_001285940.1 | 4 | 623 | NP_001272869.1 |
NM_001285946.1 | 4 | 623 | NP_001272875.1 | |
NM_001285949.1 | 4 | 623 | NP_001272878.1 | |
NM_199448.3 | 4 | 623 | NP_955519.1 | |
AK033547.1 | 4 | 565 | ||
AK150241.1 | 4 | 589 | BAE29405.1 | |
AK157221.1 | 4 | 590 | BAE34006.1 | |
AK159545.1 | 4 | 576 | BAE35171.1 | |
AK165595.1 | 4 | 573 | BAE38283.1 | |
AY382580.1 | 4 | 574 | AAR36860.1 | |
BC064130.1 | 4 | 529 | AAH64130.1 | |
BC096619.1 | 4 | 568 | AAH96619.1 |