Assay Details
Target Gene Details
Entrez Gene ID: | 11974 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit E |
Gene Aliases: |
Atp6k, Atp6v0e1, M9.2 |
Location: |
Chr.17:26676369-26699647 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp6v0e | NM_025272.2 | NP_079548.1 | ||
AK007610.1 | BAB25133.1 | |||
AK008464.1 | ||||
AK013229.1 | BAB28727.1 | |||
AK017760.1 | BAB30911.1 | |||
AK030335.1 | BAC26909.1 | |||
AK134836.1 | BAE22305.1 | |||
AK159669.1 | BAE35273.1 | |||
AK168015.1 | BAE40002.1 | |||
BC024733.1 | AAH24733.1 | |||
BY707658.1 |
More Information
Set Membership: |
Intragenic Non-exonic |