Assay Details
Target Gene Details
Entrez Gene ID: | 55979 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase, alpha) |
Gene Aliases: |
1-AGP, 1-AGPAT, AW047140 |
Location: |
Chr.17:34604278-34613449 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agpat1 | NM_001163379.1 | 4 | 773 | NP_001156851.1 |
NM_018862.3 | 4 | 525 | NP_061350.3 | |
AB005623.1 | 4 | 771 | BAA22599.1 | |
AK086281.1 | 4 | 715 | BAC39641.1 | |
BC009651.1 | 4 | 525 | AAH09651.1 |
More Information
Set Membership: |
Intragenic Exonic |