Assay Details
Target Gene Details
Entrez Gene ID: | 51789 |
Gene Name: | tyrosine kinase, non-receptor, 2 |
Gene Aliases: |
Ack, Ack-1, Ack1, Cdgip, Pyk1 |
Location: |
Chr.16:32644099-32683493 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tnk2 | NM_001110147.1 | 5 | 1012 | NP_001103617.1 |
NM_016788.3 | 5 | 1012 | NP_058068.2 | |
AF037260.1 | 5 | 795 | AAC04786.1 | |
AK087965.1 | 2 | 169 | BAC40063.1 | |
BC052421.1 | 5 | 1012 | AAH52421.1 | |
BU702226.1 | ||||
DQ666696.1 | 5 | 823 |