Assay Details
Target Gene Details
Entrez Gene ID: | 77626 |
Gene Name: | sphingomyelin phosphodiesterase 4 |
Gene Aliases: |
4122402O22Rik, mKIAA1418 |
Location: |
Chr.16:17619354-17644830 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smpd4 | NM_001164609.1 | 17 | 1807 | NP_001158081.1 |
NM_001164610.1 | 16 | 1768 | NP_001158082.1 | |
NM_001164611.1 | 16 | 1765 | NP_001158083.1 | |
NM_029945.3 | 17 | 1855 | NP_084221.2 | |
AK019466.2 | 17 | 1856 | BAB31737.1 | |
AK031029.1 | 16 | 1759 | BAC27221.1 | |
AK031685.1 | 16 | 1767 | BAC27512.1 | |
AK035892.1 | 16 | 1760 | BAC29229.1 | |
AK036055.1 | 16 | 1760 | ||
AK036380.1 | 16 | 1764 | ||
AK129354.1 | 17 | 1832 | BAC98164.1 | |
AK138720.1 | 16 | 1742 | ||
AK164234.1 | 17 | 1799 | BAE37694.1 | |
AK164456.1 | 14 | 2233 | BAE37796.1 | |
AK183804.1 | ||||
BC026767.1 | 16 | 1741 | AAH26767.1 |
More Information
Set Membership: |
Intragenic Exonic |