Assay Details
Target Gene Details
Entrez Gene ID: | 85031 |
Gene Name: | phospholipase A1 member A |
Gene Aliases: |
AA986889, Ps-pla1, Pspla1 |
Location: |
Chr.16:38396117-38433191 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla1a | NM_134102.4 | NP_598863.3 | ||
AF063498.1 | AAL55475.1 | |||
BC003470.1 | AAH03470.1 | |||
BC030670.1 | AAH30670.1 |
More Information
Set Membership: |
Intragenic Non-exonic |