Assay Details
Target Gene Details
Entrez Gene ID: | 545156 |
Gene Name: | kalirin, RhoGEF kinase |
Gene Aliases: |
2210407G14Rik, AV235988, DUET, E530005C20Rik, Gm539, Hapip, TRAD |
Location: |
Chr.16:33969073-34573291 on Build GRCm38 |
Assay Gene Location: | Within Exon 49 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kalrn | NM_177357.3 | 48 | 6875 | NP_796331.2 |
AK008844.1 | 3 | 453 | BAB25925.1 | |
AK088732.1 | 14 | 1822 | BAC40535.1 | |
AK139581.1 | 15 | 2979 | BAE24075.1 |
More Information
Set Membership: |
Intragenic Exonic |