Assay Details
Target Gene Details
Entrez Gene ID: | 22061 |
Gene Name: | transformation related protein 63 |
Gene Aliases: |
AI462811, Ket, P51/P63, P63, P73l, Tp63, Trp53rp1 |
Location: |
Chr.16:25683763-25892103 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trp63 | NM_001127259.1 | NP_001120731.1 | ||
NM_001127260.1 | NP_001120732.1 | |||
NM_001127262.1 | NP_001120734.1 | |||
NM_001127264.1 | NP_001120736.1 | |||
NM_011641.2 | NP_035771.1 | |||
AB010152.1 | BAA32432.1 | |||
AF075435.1 | AAC62640.1 | |||
AF075436.1 | AAC62641.1 | |||
AF075438.1 | AAC62643.1 | |||
AF075439.1 | AAC62644.1 | |||
AK137253.1 | BAE23286.1 | |||
BC090649.1 | AAH90649.1 |
More Information
Set Membership: |
Intragenic Non-exonic |